Description
- Methyl Butynol
Applications
- Use as inhibitor in many platinum-cured systems
Product Details
Property | Average Result |
---|---|
Appearance | Translucent |
Cure | >70 °C |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.