Description
- Two-part, white, thermally conductive silicone
- Cure may be heat accelerated
- 15:1 Mix Ratio (Part A:Part B)
Applications
- To provide heat transfer between electrical/electronic components and their heat sinks
- For use in extreme low temperature applications
- Use for adhering openings in modules and housing where grooves or other configurations require a limited flow material with moderate thermal conductivity
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 0.75 W/(mK) |
Viscosity | 75,000 cP (75,000 mPa*s) |
Cure System | Platinum |
Tensile | 275 psi (1.9 MPa) |
Appearance | White |
Cure | 30 minutes / 150 °C |
Dielectric Strength | 920 volts/mil (35.88 kV/mm) |
Durometer | 75 Type A |
Elongation | 50 % |
Mix Ratio | 15:1 |
Specific Gravity | 1.37 |
Tear | 45 ppi (7.94 kN/m) |
Work Time | 3.5 hours |
Comment | For applications requiring a broader operating temperature range. (Contains Boron Nitride) |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.