Description
- Two-part, electrically conductive RTV Silicone
- Designed for enhanced performance in extreme low and high temperatures
- 100:0.5 Mix Ratio (Base:Curing Agent)
Applications
- For RFI and EMI shielding for electrical and aerospace applications
- Excellent for formed-in-place conductive gaskets and grounding connections
- Use to adhere covers on housings or where grooves or other configurations require a non-flowable to limited flow material
Product Details
Property | Average Result |
---|---|
Cure System | Alkoxy |
Extrusion Rate | 160 g/minute |
Lap Shear | 200 psi (1.38 MPa) |
Durometer | 80 Type A |
Appearance | Gray |
Cure | 7 days / RTV |
Elongation | 90 % |
Flow Rate | 1.2 in (30.48 mm) |
Mix Ratio | 100:0.5 |
Specific Gravity | 3.41 |
Tear | 50 ppi (8.82 kN/m) |
Tensile | 250 psi (1.72 MPa) |
Volume Resistivity | 0 ohmcm |
Work Time | 3 hours |
Comment | 200 psi lap shear primed w/SP-121. Conductive at elevated temperatures. |
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.