R-2631
RFI AND EMI SHIELDING RTV SILICONE ELASTOMER
Description
  • Two-part, black, static dissipative RTV silicone
  • Based on a methyl silicone polymer
  • Supplied in a convenient 1:1 mix ratio for use with automatic mix and dispense equipment
Applications
  • For RFI and EMI shielding for industrial applications
  • Provides protection from humidity, radiation, thermal stress and mechanical stress
  • Excellent for formed-in-place conductive gaskets
  • Use to adhere covers on housings or where grooves or other configurations require a flowable to limited flow material with a moderate extrusion rate
Product Details
Property Average Result
Volume Resistivity 70 ohmcm
Cure System Platinum
Durometer 45 Type A
Tensile 615 psi (4.24 MPa)
Appearance Black
Cure 30 minutes / 150 °C
Elongation 275 %
Extrusion Rate 100 g/minute
Mix Ratio 1:1
Specific Gravity 1.08
Tear 55 ppi (9.7 kN/m)
Comment Carbon filled.
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.