Description
- Two part, Black, 1:1 Mix ratio (A:B)
- Pourable and self-leveling
- Cures at room temperature
- REACh and RoHS compliant
- Tested per UL-94 and passed V-0
Applications
- Thermally stable, flowable material that cures at room temperature
- Low viscosity allows the potting of complex geometries without the entrapment of air
- Applications include: potting for connectors, sensors, relays, power devices and other electronic components requiring protection from vibration and generally harsh environmental conditions
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 0.4 W/(mK) |
Viscosity | 2,000 cP (2,000 mPa*s) |
Cure System | Platinum |
Tensile | 525 psi (3.62 MPa) |
Appearance | Black |
Cte | |
Cure | 60 minutes / 150 °C |
Dielectric Strength | 580 volts/mil (22.62 kV/mm) |
Durometer | 50 Type A |
Elongation | 130 % |
Mix Ratio | 1:1 |
Volume Resistivity | 43,000,000,000,000 ohmcm |
Work Time | 1 hours |
Comment | 0.4 W/mk, pourable, heat resistant, tested per UL94 and passed V0. Will RTV. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.