Description
- One-part acetoxy cured silicone elastomer dispersed in Tert-Butyl Acetate
- Cures to a smooth silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance for moving, sliding, and rubbing parts
- A non-blocking topcoat for silicone surfaces that is easy to clean and prevents the accumulation of Foreign Object Debris (FOD)
Applications
- Provides a RTV, low-friction coating on cured silicone substrates
- For best results apply by spraying (dipping may also be utilized)
Product Details
Property | Average Result |
---|---|
Non Volatile Content | 7 % |
Appearance | Translucent |
Cure | RTV |
Cure Rate | |
Cure System | Acetoxy |
Comment | One Part and cures at room temperature. Acetic acid cure by-product therefore not recommended where corrosion is of concern. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.