Description
- A clear, tacky silicone gel
- A medium viscosity liquid that cures to a soft, high penetration silicone
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for applications requiring a clear, tacky silicone gel
- Ideal for casting, calendering, backfilling, and dampening
- Consider for transdermal applications
Product Details
Property | Average Result |
---|---|
Penetration | 5 mm |
Viscosity | 15,300 cP (15,300 mPa*s) |
Mix Ratio | 1:1 |
Appearance | Translucent |
Cure | 60 °C |
Cure Rate | 3 ppm/°C |
Cure System | Platinum |
Tack | 5.8 psi (0.04 MPa) |
Work Time | 30 minutes |
Comment | Medium Viscosity, Soft Gel |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.