Description
- Two-part, translucent, pourable silicone elastomer
- Offers high tear strength and excellent physical properties
- Cures with heat via addition-cure chemistry
- 10:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property | Average Result |
---|---|
Durometer | 40 Type A |
Viscosity | 73,400 cP (73,400 mPa*s) |
Work Time | 5 hours |
Tensile | 875 psi (6.03 MPa) |
Appearance | Translucent |
Cure | 5 minutes / 177 °C |
Cure System | Platinum |
Elongation | 360 % |
Mix Ratio | 10:1 |
Post Cure | 1 minute / 150 °C |
Specific Gravity | 1.09 |
Stress at Strain | 480 psi / 200 % (3.31 MPa / 200 %) |
Tear | 150 ppi (26.46 kN/m) |
Comment | Flowable, Medium Durometer |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.