Premium Care Line
MED-4042
HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • A unique three-part system, VersaSil3 offers the flexibility of adjustable cure rate and table life for various fabrication requirements
  • High tear strength, wide processing parameters, and translucent, non-tacky surfaces
  • Developed to be compounded with the inhibitor CAT-40 and the platinum catalyst CAT-55. Advantages include: no volatile by-products, lower cure temperature, and optional post-cure
  • 100: 1.0: 1.0 Mix Ratio
Applications
  • For mass production with maximum flexibility
  • For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property Average Result
Durometer 40 Type A
Tensile 1,500 psi (10.34 MPa)
Elongation 1000 %
Tear 160 ppi (28.22 kN/m)
Cure 10 minutes / 116 °C
Cure Rate Scorch 0.8 minutes
Cure Rate T90 2.45 minutes
Mix Ratio 100:1:1
Special Feature VersaSil
Specific Gravity 1.11
Stress at Strain 230 psi / 200 % (1.59 MPa / 200 %)
Work Time 2 hours
Comment Can be Cured via Platinum or Peroxide
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.