Description
- Specially formulated primer
- Easy to use, air-drying, one-part primer designed for use with platinum-cured systems
- One-component primer supplied in Isopropyl Alcohol requires no mixing
- Flash Point (TCC) of 12°C (53°F)
- Convenient container sizes yield less waste
- Lower Volatile Organic Compound (VOC) content
Applications
- Improves adhesion of silicones to various substrates including: metals (such as stainless steel, steel, copper and aluminum), ceramics, rigid plastics, and other silicone materials
NuSil Technology’s MED-166 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Cure | 1 hours / RTV |
Mix Ratio | 1 PART |
Non Volatile Content | 6.5 % |
Specific Gravity | 0.8 |
Appearance | Translucent |
Solvent | IPA |
Comment | Dispersed in Isopropyl Alcohol, compatible with acrylics. |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.