LS-3246
ULTRA-CLEAR ENCAPSULATION GEL
Description
  • A two-part, encapsulation gel
  • Extended work time
  • 1.46 refractive index for matching silica
  • 1:1 Mix Ratio (Part A:B)
Applications
  • For protection of sensitive photonics assemblies from mechanical shock, thermal shock, dust, and ambient atmosphere
  • Extended work time allows voids in complex assemblies to fill in and permits time for any trapped air bubbles to float to the fluid surface and escape
  • For applications requiring an operating temperature range of -40°C to 200°C
  • (-40°F to 392°F) or for soldering operations up to 260°C (500°F) for 1 to 2 minutes
Product Details
Property Average Result
Viscosity 1,000 cP (1,000 MPa*s)
Work Time 8 hours
Appearance Clear
Cure 1 minutes / 65 °C
Cure System Platinum
Durometer 10 Type 00
Mix Ratio 1:1
Operating Temperature
Refractive Index 1.46
Specific Gravity 1.04
Comment Firm Gel;Index Matches 1.46 RI such as fused silica. Will RTV within 24 hrs. 5 hr. minimum potlife based on 2X initial viscosity.
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.