Description
- Strong adhesion to other silicones
- Dispersed in Xylene
- Accelerated cure with introduction of heat
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Ideal for use as a marking ink for silicone parts and other
- components on which the coating must maintain long-term
- stability
- Use in applications requiring a rapid cure schedule
- Provides high opacity for single print applications
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 0.68 W/(mK) |
Cure System | Non-Curing |
Appearance | White |
Flow Rate | 0.2 in (5.08 mm) |
Specific Gravity | 2.26 |
Comment | Versatile, thermally conductive grease where thin bondlines or a compressible grease is needed. Non-curing. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.