Description
- Two-part, white, thermally conductive, low viscosity silicone elastomer
- Cures with the addition of heat
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring a low volatility, conformal, thermally conductive silicone
- To provide heat transfer between electrical/electronic components and their heat sinks
- Use to adhere integrated circuit substrates, base plates, heat sinks or where grooves or other configurations require a limited flow material
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 0.95 W/(mK) |
Viscosity | 36,000 cP (36,000 mPa*s) |
Cure System | Platinum |
Tensile | 180 psi (1.24 MPa) |
Appearance | White |
Cure | 14 minutes / 150 °C |
Dielectric Strength | 465 volts/mil (18.14 kV/mm) |
Durometer | 65 Type A |
Elongation | 50 % |
Ionic Content Cl | <5 ppm |
Ionic Content K | <3 ppm |
Ionic Content Na | <5 ppm |
Lap Shear | 120 psi (0.83 MPa) |
Mix Ratio | 1:1 |
Specific Gravity | 2.34 |
Tack Free Time | 13 hours |
Volatile Content | 0.35 % |
Volume Resistivity | 34,000,000,000,000 ohmcm |
Comment | Recommended for bondlines 5 micron or greater. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.