Description
- One-Part, translucent, non-slumping RTV silicone
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- As a sealing, caulking, adhesive or potting material in electronic and space applications requiring minimal outgassing to avoid condensation in sensitive devices
- For bonding and sealing in applications such as overhead or vertical joints that require a non-slumping and one-part material
Product Details
Property | Average Result |
---|---|
Cure System | Oxime |
Extrusion Rate | 25 g/minute |
Lap Shear | 350 psi (2.41 MPa) |
Durometer | 45 Type A |
Appearance | Translucent |
Cure | 7 days / RTV |
Elongation | 400 % |
Mix Ratio | 1 PART |
Specific Gravity | 1.1 |
Tack Free Time | 15 minutes |
Tensile | 800 psi (5.52 MPa) |
Comment | Non-Slump |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.