Orders LinkContactUsLinkComponent
Masterflex I/P Tubing Options | Avantor
Masterflex I/P Tubing Options
Achieve top performance from your Masterflex I/P pump heads when you use precision-extruded tubing. Tubing is manufactured to tight tolerances that match our I/P pump heads. Choose from precision tubing or high-performance precision tubing. High-performance precision tubing has a thicker wall that improves pressure generation, suction lift, ability to pump viscous fluids, and tubing life. Choose from over a dozen different formulations.
Precision Tubing Specifications
Tubing sizeI/P 26I/P 73I/P 82
Inside diameter (nominal)0.25" (6.4 mm)0.37" (9.5 mm)0.5" (12.7 mm)
Hose barb size (nominal)1/4" (6.4 mm)3/8" (9.5 mm)1/2" (12.7 mm)
Flow range (approximate) with 20 to 650 rpm drive0.12 to 4 L/min
(0.03 to 1.1 GPM)
0.2 to 8 L/min
(0.05 to 2.1 GPM)
0.4 to 13 L/min
(0.1 to 3.5 GPM)
Maximum pressure, continuous**25 psig (1.7 bar)10 psig (0.7 bar)
Maximum pressure, intermittent**40 psig (2.7 bar)20 psig (1.4 bar)
Maximum vacuum**26" Hg (660 mm Hg)20" Hg (510 mm Hg)
Suction lift29 ft H2O (8.8 m H2O)23 ft H2O (7.0 m H2O)
High-Performance Precision Tubing Specifications
Tubing sizeI/P 70I/P 88I/P 89
Inside diameter (nominal)0.37" (6.4 mm)0.5" (12.7 mm)0.62"
(15.88 mm)
Hose barb size (nominal)3/8" (9.5 mm)1/2" (12.7 mm)5/8" (15.88 mm)
Flow range (approximate) with 20 to 650 rpm drive0.24 to 8 L/min
(0.06 to 10 GPM)
0.5 to 15 L/min
(0.05 to 2.1 GPM)
0.52 to 17 L/min
(0.14 to 4.5 GPM)
Maximum pressure, continuous**25 psig (1.7 bar)20 psig (1.4 bar)15 psig (1.0 bar)
Maximum pressure, intermittent**40 psig (2.7 bar)35 psig (1.4 bar)25 psig (1.4 bar)
Maximum vacuum**26" Hg (660 mm Hg)24" Hg (610 mm Hg)
Suction lift29 ft H2O (8.8 m H2O)27 ft H2O (8.2 m H2O)
**Actual performance varies depending on tubing formulation—values shown are for firm tubing.