Orders LinkContactUsLinkComponent
C-FLEX (50 A) Tubing | Avantor
C-FLEX (50 A) Tubing
Advantages:
  • Physical properties similar to silicone with chemical compatibility of TYGON®.
  • Inexpensive.
  • Excellent biocompatibility.
  • Smooth bore.
  • Nontoxic, no leachable plasticizers.
  • Lower gas permeability than silicone.
  • Use with many acids and alkalies.
Limitations:
  • Not recommended for use with oils.
  • Moderate pumping life.
Application Suitability:
Acids:Good
Alkalies:Good
Organic solvents:Not recommended
Pressure:Fair
Vacuum:Good
Viscous fluids:Fair
Sterile fluids:Excellent
Physical Characteristics and Compostion:
  • Thermoplastic elastomer.
  • Styrene-ethylene-butylene modified block copolymer with silicone oil.
  • Excellent tensile and tear strength.
  • Soft material.
  • Opaque, white.
Temperature Range:
-100 to 275°F (-73 to 135°C)
Meets Classifications:
  • USP Class VI
  • FDA 21 CFR 177.2600
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2:-
H2:-
O2:150
N2:-
Cleaning/Sterilization:
  • Sterilize by ethylene oxide (ETO), autoclave or gamma irradiation.