Orders LinkContactUsLinkComponent
PHARMED Tubing | Avantor
PHARMED Tubing
Advantages:
  • Great for tissue and cell work nontoxic and nonhemolytic
  • Long service life minimizes risk of fluid exposure; reduces tubing costs and pump downtime
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Heat sealable, bondable, and formable
  • Extremely low gas permeability
Limitations:
  • Potential leaching of USP mineral oil or blend materialApplication Suitability:
Application Suitability:
Acids:Good
Alkalies:Good
Organic solvents:Not recommended
Pressure:Good
Vacuum:Excellent
Viscous fluids:Good
Sterile fluids:Excellent
Physical Characteristics and Compostion:
  • Thermoplastic elastomer.
  • Polypropylene-based material with USP mineral oil.
  • Excellent tensile strength.
  • Firm (stiff) material.
  • Opaque, beige.
Temperature Range:
-75 to 275°F (-59 to 135°C)
Meets Classifications:
  • USP Class VI
  • FDA 21 CFR 177.2600
  • NSF listed (Standard 51)
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2:1200
H2:-
O2:200
N2:80
Cleaning/Sterilization:
  • Sterilize by ethylene oxide (ETO), autoclave or gamma irradiation up to 2.5 Mrad.
  • Repeated autoclaving will not affect overall life.