Orders LinkContactUsLinkComponent
mflx-ls-mastersense-fill-finish-pump-system-ip66-1920x760.jpg
Masterflex® L/S® MasterSense™ Fill/Finish Pump System with Masterflex® Ultrapharm™ Pump Head
Accuracy is critical when every drop counts
Masterflex® L/S® MasterSense™ Fill/Finish Pump System with Masterflex® UltraPharm™ Pump Head
Shop now
Masterflex® L/S® MasterSense™ Fill/Finish Pump System with Masterflex® Ultrapharm™ Pump Head | Avantor
Features that make your process effortless
ultrapharm-bundle-callouts.png
  1. Perfect for sanitary environments—316 polished stainless steel drive housing, IP66 and NEMA 4X rated for washdown
  2. Easy to read and operate touch screen - even while wearing gloves. Create, save, and recall pump programs via intuitive menus.
  3. EtherNet/IP, Profibus, Wi-Fi, MasterflexLive enabled, and analog remote I/O means connectivity and control options that suit your needs.
  4. 250 rpm max speed; dosing volumes from <0.5 mL to >500 mL
  5. Whisper quiet operation reduces extra noise on the floor or in the lab
  6. 316 stainless steel pump head is autoclavable; tested to 10 cycles without any loss of flow or accuracy performance
  7. Offset rollers and planetary gearing ensure dosing accuracy (±0.5%), low pulsation, and long tubing life
    Easy on tubing— reduces drag on tubing for longer tubing life and reduced risk of premature failure.
  8. Masterflex double-Y tube sets ensure proper retention, repeatability, and accuracy
    SHOP DOUBLE Y-TUBING
image_150x249.jpg
Continuous Mode
mastersense-bt-biopharma-time-sm.png
Time Mode
mastersense-bt-biopharma-volume-sm.png
Volume Mode
mastersense-bt-biopharma-ethernet-sm.png
Remote Mode
mastersense-bt-biopharma-tubcal-sm.png
Tube Calibration
mastersense-ls-tiereduser-sm.png
User Access
mflx-planetary-drive.jpg
Benefits of a Pump Head with Planetary Drive System
With the planetary drive system, each roller is directly driven by the sun wheel. This mechanism prevents axial push-pull friction on the tubing—resulting in longer tubing life, lower pulsation, and high repeatability.
This pump head produces gentle, low pulse, and highly accurate dosing and flow to ensure optimal integrity and uniformity. Designed with offset rollers, the pump head provides linear flow throughout the flow range. While one roller is drawing fluid through one leg of the double-Y tube set, the other roller is pushing fluid through the other leg. Also, dual channels allow for a lower rpm in order to achieve “single-channel” flow rates. A lower rpm means reduced shear forces to maintain fluid and molecular integrity.
touchscreen-closeup.jpg
Set up is a breeze
Intuitive touch screen interface, even wearing gloves
  • Easy to set up and program
  • 5" capacitive TFT/RGB touch screen with a clear, wide viewing angle
  • Adjust parameters with a simple touch—even with gloves on
  • Operational modes include continuous run, volume dispense, and time dispense
  • Batch control and interval/off time in Volume and Time modes
  • Antidrip and speed ramping available in Volume mode—ensure accuracy and minimize sample loss
  • Easily save and recall up to 50 commonly used protocols, each with a unique name
  • Know when you hit your mark—graphical indicator lets you know when loaded tubing is optimally calibrated
Advanced Connectivity and Control Options
Monitor from anywhere
connectivity-img-diagram-LSultrapharm.png
MasterflexLive
  • MasterflexLive is our secure, cloud-enabled platform for real-time remote monitoring of all Masterflex pump parameters
  • Monitor critical processes running 24/7 without the need for onsite personnel
  • Push notifications alert you to changes in operating conditions and error messages
Learn more
Flexible Connectivity
  • Your choice of connectivity options— EtherNet/IP, Profibus, or MasterflexLive®
  • Three-tier access to pump functionality—use this option to protect your process by controlling who can operate the pump and at what level
  • Over-the-air firmware updates—automatically allows you to take full advantage of future enhancements
  • Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive
The Perfect Pump for High Precision Fill/Finish Applications
biopharma.jpg
Non-contaminating, low shear, and noninvasive makes this pump perfect for Biopharma and healthcare.
vacc-productionline.jpg
Three-tier access and the ability to save and recall programs make your fluidic process secure and accurate.
masterflex-tubing-263x183.jpg
Our range of tubing formulations ensures the fluid compatibility and the regulatory approvals you need.
water-treatment.jpg
Precision manufactured pump tubing gives you accuracy and reliability for critical process applications.
Complete Specifications & Product Details
 Masterflex L/S MasterSense Fill/Finish pump systemMasterflex L/S MasterSense Fill/Finish pump system
Item #MFLX77976-40MFLX77976-42
Flow Range0.007 to 635 mL/min0.193 to 998 mL/min
Masterflex Ultrapharm Pump Headfor Precision Tubing with Open-Head Sensor, stainless steel housing/rotorfor High-Performance Precision Tubing with Open-Head Sensor, stainless steel housing/rotor
Tubing sizes acceptedDouble-Y L/S® 13, 14, 16, 25, 17Double-Y L/S® 15, 24, 35
Pump Specifications
Agency ApprovalsUL, cUL, CE
Speed control accuracy (resolution)±0.1% (0.1 rpm at 250 rpm)
Warranty2 Year
MasterflexLiveReal-time remote monitoring of all pump parameters. Push notifications alert you to operating conditions and error messages, allowing you to monitor critical pump processes 24/7—no need to be onsite.
EtherNet/IP or
Profibus
Interface directly with either Ethernet/IP or Profibus based process control network(s) (using PLC, SCADA or similar); enables control of all pump parameters via network interface.
Number of rollers8
ReversibleYes
Display5” capacitive TFT/RGB touch screen with in-plane switching (wide viewing angle); 800 x 480 pixels
Digital DispensingYes
External control - input0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; start/stop, direction (CW/CCW), prime via contact closure
External control - output0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Tach output (open collector), Motor running, Open head, Local/Remote, General alarm
MotorBrushless DC with encoder; continuous duty; reversible; 1/10 hp (75 W)
Open-head sensorYes; extension cable included
Housing materials316 Stainless Steel
Enclosure ratingIP66 / NEMA 4X
Dimensions (L x W x H)13" x 9.3" x 11" (33 x 23.6 x 27.9 cm)
Shipping weight40.75 lb (18.5 kg)
Power90 to 260 VAC, 50/60 Hz