ContactUsLinkComponent
Product with code:39208459 not found
Product with code:39209011 not found
Product with code:39209009 not found
masterflexlive1920x760.jpg
MasterflexLive® Remote Peristaltic Pump Monitoring System
Monitor critical pump processes 24/7 with MasterflexLive
Get the most out of your MasterflexLive® apps, answer questions, and solve problems.
MasterflexLive User Guide
MasterflexLive remote pump monitoring system makes it simple to get all the pump parameters including speed, flow rate, dispense volume, and more in real-time, from any location.
masterflex-live-logo-38h.png
Sign up in minutes with 2 easy steps
Simple setup
  • Register a new MasterflexLive account
  • Enter your device ID number (MAC address) and model code
Already set up? Sign In
Register now
Features and benefits of MasterflexLive remote pump monitoring
You simply can’t be everywhere at once. With MasterflexLive® you can get real-time monitoring of your Masterflex pump system.  
Push notifications alert you to operating conditions and errors, so you’re never out of the loop. Your MasterflexLive remote pump monitoring system means you can keep track of what’s going on even when you’re offsite.   
MasterflexLive offers free activation of your pump, plus the following benefits: 
  • Real-time remote monitoring
  • Pump status notifications
  • Monitoring of all pump parameters
  • Unlimited pumps per account
  • Multiple user levels with password security
  • Front sensor warning (open head and leak detection)
Masterflex Cloud-Enabled Systems
We can help you set up a cloud-based fluid handling system for your specific needs