Orders LinkContactUsLinkComponent
ECMs & Coatings | Avantor
ECMs & Coatings
Featured Solutions
coating1.jpg
Cells behave better on Corning Matrigel matrix - the original, trusted extracellular matrix (ECM). The natural ECM-based hydrogel is among the most widely used models for 2D and 3D cell culture in vitro.
Additional Resources
Matrigel® Matrix for organoid culture
greengrowdox.jpg
UPM Biomedicals develops and supplies innovative and sustainable wood-based biomedical products. GrowDex® is a nanofibrilar cellulose hydrogel for 3D cell culturing.
Additional Resources
More information