Orders LinkContactUsLinkComponent
Cell Counting | Avantor
Cell Counting
EVE.jpg
EVE is a benchtop automatic cell counter designed to measure cell count and viability accurately and precisely. It uses the standard trypan blue technique
Additional Resources
Learn more about EVE™
EVEPlus.jpg
EVE PLUS is a compact image-based automated cell counter that uses disposable slides to most accurately determine eukaryotic cell titers in less than 1 second.
Additional Resources
Learn more about EVE™ Plus
ADAM-MC2.jpg
ADAM-MC2 is an automated fluorescence cell counter that performs cell counting and viability measurements at highest accuracy capable of replacing complex flow cytometry workflows. Combined with the disposable micro-fluidic Accuchip, the operation is extremely simple and cost-effective.
Additional Resources
Learn more about ADAM-MC2
C-Slide-Cel.jpg
C-Slide Cell counting chamber slides for use with automated cell counter (Countess and Countess II).
Additional Resources
Learn more about C-Slide