Orders LinkContactUsLinkComponent
mastersense-stainless-steel-pump-drive-no-icon-1920x760.jpg
Masterflex® L/S® MasterSense™ BioPharma Process Drives
Reimage bioprocessing
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Masterflex® L/S® MasterSense™ Stainless Steel Digital Pump Drives | Avantor
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
MasterSense_monitors_your_media_so_you_dont_have_t.png
Advanced Connectivity and Control Options
Monitor from anywhere
ls-washdown-connectivity-img-diagram-LS.png
MasterflexLive
  • MasterflexLive is our secure, cloud-enabled platform for real-time remote monitoring of all Masterflex pump parameters
  • Monitor critical processes running 24/7 without the need for onsite personnel
  • Push notifications alert you to changes in operating conditions and error messages
Learn more
Flexible Connectivity
  • Your choice of connectivity options—MasterflexLive®, EtherNet/IP, or Profibus
  • Three-tier access to pump functionality—use this option to protect your process by controlling who can operate the pump and at what level
  • Over-the-air firmware updates—automatically allows you to take full advantage of future enhancements
  • Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive
Complete your pump system
Choose from 9 pump heads and 20+ tubing formulations to create the perfect pump for your process
Complete Specifications & Product Details
Types of pump heads accepted
Types of pump heads accepted)
Masterflex L/S MasterSense Drive 600 RPM (up to 4 pump heads accepted)Masterflex L/S MasterSense Process Drive 600 RPM with Easy-Load pump head BundleMasterflex L/S MasterSense Process Drive 600 RPM with Easy-Load II pump head Bundle
600 RPM
(up to 2 pump heads accepted)
600 RPM
Easy-Load
Complete System
600 RPM
Easy-Load II
Complete System
Flow Range0.006 to 3400 mL/min0.08 to 480 mL/min0.28 to 1700 mL/min
With EtherNet/IP and MasterflexLiveMFLX07576-10MFLX77976-00MFLX77976-02
With Profibus and MasterflexLiveMFLX07576-12MFLX77976-10MFLX77976-12
Drive Specifications
Agency ApprovalsUL, cUL, CE
Warranty2 Year
MasterflexLiveReal-time remote monitoring of all pump parameters. Push notifications alert you to operating conditions and error messages, allowing you to monitor critical pump processes 24/7—no need to be onsite.
EtherNet/IP or
Profibus
Interface directly with either Ethernet/IP or Profibus based process control network(s) (using PLC, SCADA or similar); enables control of all pump parameters via network interface.
Speed control accuracy (resolution)±0.1% (0.1 rpm at 600 rpm)
ReversibleYes
Display5” capacitive TFT/RGB touch screen with in-plane switching (wide viewing angle); 800 x 480 pixels
Digital DispensingYes
External control - input0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; start/stop, direction (CW/CCW), prime via contact closure
External control - output0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Tach output (open collector), Motor running, Open head, Local/Remote, General alarm
MotorBrushless DC with encoder; continuous duty; reversible; 1/10 hp (75 W)
Open-head sensorYes; extension cable included
Housing materials316 Stainless Steel
Enclosure ratingIP66 / NEMA 4X
Dimensions (L x W x H)13" x 9.3" x 11" (33 x 23.6 x 27.9 cm)
Shipping weight28.75 lb (11.794 kg)
Power90 to 260 VAC, 50/60 Hz
biopharma.jpg
Non-contaminating, low shear, and noninvasive makes this pump perfect for Biopharma and healthcare.
vacc-productionline.jpg
Three-tier access and the ability to save and recall programs make your fluidic process secure and accurate.
blue-chem-liquid.jpg
Our range of tubing formulations ensures the fluid compatibility and the regulatory approvals you need.
water-treatment.jpg
Precision manufactured pump tubing gives you accuracy and reliability for critical process applications.
touchscreen-closeup.jpg
Set up is a breeze
Intuitive touch screen interface, even wearing gloves
  • Easy to set up and program
  • 5" capacitive TFT/RGB touch screen with a clear, wide viewing angle
  • Adjust parameters with a simple touch—even with gloves on
  • Operational modes include continuous run, volume dispense, and time dispense
  • Batch control and interval/off time in Volume and Time modes
  • Antidrip and speed ramping available in Volume mode—ensure accuracy and minimize sample loss
  • Easily save and recall up to 50 commonly used protocols, each with a unique name
  • Know when you hit your mark—graphical indicator lets you know when loaded tubing is optimally calibrated
Take your process to the next level
MasterSense_BioPharma_Process_Drives.png
  1. Perfect for sanitary environments—316 polished stainless steel housing IP66 NEMA 4X rated for washdown
  2. Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
  3. Intuitive digital touchscreen with optimized menu navigation to store and recall standard protocols ensuring continuity-even with wet gloves on!
  4. Over-the-air firmware updates ensure optimization of your pump system without moving the system while MasterflexLive® cloud storage system securely stores, monitors and generates reports for all Masterflex pump systems
  5. Powerful performance in a small footprint. Variable flow up to 3400 mL/min (600 rpm). Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio
    SHOP DRIVE OPTIONS
  6. Qualitative tube calibration ensures tubing is calibrated precisely
    SHOP L/S PUMP TUBING
  7. Convenient open-head sensor and antidrip features. Accepts all L/S pump heads.
    SHOP L/S PUMP HEADS
image_150x249.jpg
Continuous Mode
mastersense-bt-biopharma-time-sm.png
Time Mode
mastersense-bt-biopharma-volume-sm.png
Volume Mode
mastersense-bt-biopharma-ethernet-sm.png
Remote Mode
mastersense-bt-biopharma-tubcal-sm.png
Tube Calibration
mastersense-ls-tiereduser-sm.png
User Access