Orders LinkContactUsLinkComponent
masterflex-ls-mastersense-drives-1920x760.jpg
Masterflex® L/S® MasterSense™ Drives
Reimage fluid handling
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Masterflex® L/S® MasterSense™ Drives | Avantor
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
MasterSense_monitors_your_media_so_you_dont_have_t.png
Monitor your process remotely and stored programs securely
connectivity-img-diagram.png
MasterflexLive
  • MasterflexLive is our secure, cloud-enabled platform for real-time remote monitoring of all Masterflex pump parameters
  • Monitor critical processes running 24/7 without the need for onsite personnel
  • Push notifications alert you to changes in operating conditions and error messages
Learn more
Flexible Connectivity
  • Your choice of connectivity options—MasterflexLive®, EtherNet/IP, or Profibus
  • Three-tier access to pump functionality—use this option to protect your process by controlling who can operate the pump and at what level
  • Over-the-air firmware updates—automatically allows you to take full advantage of future enhancements
  • Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive
Complete your pump system
Choose from 8 pump heads and 20+ tubing formulations to create the perfect pump for your process
Choose the drive or complete pump that is right for you
Masterflex L/S MasterSense Drive 100 RPM (up to 4 pump heads accepted)Masterflex L/S MasterSense Drive 600 RPM (up to 4 pump heads accepted)Masterflex L/S MasterSense Drive 600 RPM with Easy-Load pump head BundleMasterflex L/S MasterSense Drive 600 RPM with Easy-Load II pump head Bundle
100 RPM
(up to 4 pump heads accepted)
600 RPM
(up to 2 pump heads accepted)
600 RPM
Easy-Load
Complete System
600 RPM
Easy-Load II
Complete System
Flow Range0.001 to 580 mL/min0.006 to 3400 mL/min0.08 to 480 mL/min0.28 to 1700 mL/min
With EtherNet/IP and MasterflexLiveMFLX07526-20MFLX07526-10MFLX77920-40MFLX77920-42
With Profibus and MasterflexLiveMFLX07526-22MFLX07526-12MFLX77920-50MFLX77920-52
Drive Specifications
Agency ApprovalsUL, cUL, CE
Warranty2 Year
MasterflexLiveReal-time remote monitoring of all pump parameters. Push notifications alert you to operating conditions and error messages, allowing you to monitor critical pump processes 24/7—no need to be onsite.
EtherNet/IP or
Profibus
Interface directly with either Ethernet/IP or Profibus based process control network(s) (using PLC, SCADA or similar); enables control of all pump parameters via network interface.
Speed control accuracy (resolution)±0.1% (0.1 rpm at 600 rpm; 0.01 rpm at 100 rpm)
ReversibleYes
Display5” capacitive TFT/RGB touch screen with in-plane switching (wide viewing angle); 800 x 480 pixels
Digital DispensingYes
External control - input0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; start/stop, direction (CW/CCW), prime via contact closure
External control - output0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Tach output (open collector), Motor running, Open head, Local/Remote, General alarm
MotorBrushless DC with encoder; continuous duty; reversible; 1/10 hp (75 W)
Open-head sensorYes; extension cable included
Housing materialsABS plastic housing, painted aluminum chassis, painted aluminum bezel
Enclosure ratingIP33
Dimensions (L x W x H)10-1/4" x 7-5/8" x 8-1/2" (26 x 19.4 x 21.6 cm)
Shipping weight11 lb (5 kg)
Power90 to 260 VAC, 50/60 Hz
biopharma.jpg
Non-contaminating, low shear, and noninvasive makes this pump perfect for Biopharma and healthcare. 
vacc-productionline.jpg
Three-tier access and the ability to save and recall programs make your fluidic process secure and accurate.
blue-chem-liquid.jpg
Easily handles abrasive and corrosive fluids found in many chemical and mining applications.
water-treatment.jpg
Long life tubing prevents chemical leaks and elimination of vapor-lock, important for water treatment processes.
touchscreen-closeup.jpg
Skip reading the operation manual
Intuitive touch screen interface, even wearing gloves
  • Easy to set up and program
  • 5" capacitive TFT/RGB touch screen with a clear, wide viewing angle
  • Adjust parameters with a simple touch—even with gloves on
  • Operational modes include continuous run, volume dispense, and time dispense
  • Batch control and interval/off time in Volume and Time modes
  • Antidrip and speed ramping available in Volume mode—ensure accuracy and minimize sample loss
  • Easily save and recall up to 50 commonly used protocols, each with a unique name
  • Know when you hit your mark—graphical indicator lets you know when loaded tubing is optimally calibrated
image_150x249.jpg
Continuous Mode
mastersense-bt-biopharma-time-sm.png
Time Mode
mastersense-bt-biopharma-volume-sm.png
Volume Mode
mastersense-bt-biopharma-ethernet-sm.png
Remote Mode
mastersense-bt-biopharma-tubcal-sm.png
Tube Calibration
mastersense-ls-tiereduser-sm.png
User Access
Features that make your process effortless
MasterSense_Drives.png
  1. Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
  2. Intuitive digital touchscreen with optimized menu navigation to store and recall standard protocols ensuring continuity - even with wet gloves on!
  3. Over-the-air firmware updates ensure optimization of your pump system without moving the system
  4. MasterflexLive cloud storage system securely stores, monitors and generates reports for all Masterflex pump systems
  5. Powerful performance in a small footprint. Variable flow up to 3400 mL/min (600 rpm). Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio
    SHOP DRIVE OPTIONS
  6. Qualitative tube calibration ensures tubing is calibrated precisely
    SHOP L/S PUMP TUBING
  7. Convenient open-head sensor and antidrip features. Accepts all L/S pump heads.
    SHOP L/S PUMP HEADS