Orders LinkContactUsLinkComponent
SwTechnologiesHeader.jpg
SW Technologies
SW® invests in manufacturing, glove performance, and hand health technologies to provide premium products that protect and enhance worker performance.
SW Technologies | Avantor
MANUFACTURING – SW has focused on developing new manufacturing technologies to drive category innovation and improve worker performance.
HAND HEALTH – SW proprietary technologies that improve hand health and counter the effects associated with wearing hand protection.
GLOVE PERFORMANCE – SW has developed a number of new technologies that have transformed the single-use glove category – from simply protecting hands to enhancing comfort, enabling better performance, to protecting the environment.
1TechnologiesCard.jpg
EnerGel™
Hydrates and rejuvenates hands for all-day comfort. New patented additive clinically proven to improve hand health.
2TechnologiesCard.jpg
pH Natural®
Actively maintains skin pH at natural levels to reduce skin issues. Proprietary coating helps keep pH levels at 5.5, reducing microbial growth.
3TechnologiesCard.jpg
Dry Core™
Naturally pulls sweat away from the skin for healthier hands. Moisture-wicking system makes for a dry, comfortable environment.
4TechnologiesCard.jpg
DriTek®
Moisture-wicking flock-lined system for drier, cooler, more comfortable hands. Naturally manages sweat for healthier hands and easy donning and doffing.
5TechnologiesCard.jpg
EcoTek®
EcoTek® is a proprietary nitrile treatment that accelerates waste reduction in landfills. SW® nitrile gloves with EcoTek reduce waste by 92.6% in only 2 years and 6 months under test method ASTM D5526-12, a standard test method for determining anaerobic biodegradation of plastics in accelerated landfill conditions. Current biodegradation results do not predict future results.
6TechnologiesCard.jpg
TracTek®
Proprietary matrixed pattern that provides dramatic control. Designed for maximum surface-to-surface contact, providing enhanced grip in all conditions.
7TechnologiesCard.jpg
Breach Alert™
Dual color, 2-ply construction technology provides immediate visual breach detection. Two distinct polymer layers add durability and dramatically improve safety.
9TechnologiesCard.jpg
AxiFybr®
Proprietary molecularly-engineered yarn that dramatically enhances cut resistance. Provides the highest level cut protection with high glove comfort and dexterity.