Orders LinkContactUsLinkComponent
363165FeatureCard.jpg
Sampling Accessories
The right accessories will help you to round-off your sampling process. This chapter shows a small selection of useful tools to optimise your sampling.
Sampling accessories | Avantor
363165CC1.jpg
Close-it labels
Stick firmly to most materials, even to surfaces that are slightly moist or coated with powder. Ideal for QC and repairing damaged packaging.
363165CC2.jpg
Seals
Useable as original seals and protection from sample manipulation. The initial opening of the packaging is made obvious and irrevocably visible.
363165CC3.jpg
Sampling bags and tubes
363165CC4.jpg
Cleaning brushes
Help to clean different kind of sampling lances thoroughly. Equipped with strong efficient bristles.
363165CC5.jpg
Snowpack dry ice machine
Enables you to obtain a -79 °C dry ice tablet. Ideal for lowering temperatures within cooling baths or boxes for sample storage or transport.
363165CC6new.jpg
Accessories for zone samplers
These special accessories help to collect and release samples.