Order Entry
Belgium
Orders LinkContactUsLinkComponent
Security seals, SampleSafe

Security seals, SampleSafe

SampleSafe seal, PP, red, (Ø×L) 2×260 mm
Catalog # 331-0057
Supplier:   BURKLE
Security seals, SampleSafe
Catalog # 331-0057
Supplier:  BURKLE
Security seals, SampleSafe
Security seals, SampleSafe

SampleSafe seal, PP, red, (Ø×L) 2×260 mm

Price
133,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

SampleSafe seals are made of PP and are uniquely identified by a 9-digit serial number.

  • Serial numbering for clear identification
  • Stainless steel safety spring provides tamper protection

The seal ring (Ø 2 mm) is able to fit through small eyelets and slits. The tape is secured by a stainless steel spring with one-way tightening mechanism, which ensures a tamper-evident seal.

Specifications
  • Description:
    SampleSafe seal, PP, red, (Ø×L) 2×260 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.