Orders LinkContactUsLinkComponent
Redefinin-Microscopy-Feature.jpg
Redefining Microscopy For Life Science Research
Discover Echo has been built around a singular vision to rethink traditional microscope design. Every aspect of workflow, usability, and performance have been pushed to create a more modern and powerful product.
71% of research labs today own both UPRIGHT and INVERTED microscopes.
Echo has developed the world's first hybrid microscopes that can easily convert between either configuration. There is no other microscope in the market that can provide this.
We have also replaced eyepieces with ultra- high resolution touch displays to create an unparalleled experience.
Request a call or demo
ECHO - Redefining Microscopy | Avantor
Rebel
Quick-reliable-540.jpg
Quick, reliable, high quality images, at your fingertips
A workhorse microscope for brightfield/tissue culture.
Upright and inverted configuration Imaging modes: BF-PH-POL 
5 position intelligent nosepiece.
DISCOVER MORE
Revolve
revolve-4k-540.jpg
Discover, capture, share, and publish - a microscope your entire lab will use
Powerful, advanced research level microscope
Upright and inverted configuration Imaging modes: FL, BF, PH, DF, POL
6 Position intelligent nosepiece
5 Channel motorised fluorescence
Z-Stack available
Lite automation
DISCOVER MORE
Revolution
Revolution-540.jpg
The automated microscope for life science research
Fully automated, high performance imaging
Upright and inverted configurations Imaging modes: FL, BF, PH, DF, POL
6 Position intelligent nosepiece
5 Channel motorised fluorescence
Z-Stack included
Auto focus
Full automation
DISCOVER MORE