Order Entry
Austria
ContactUsLinkComponent
 
ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright/Inverted)
 
undefined
ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright/Inverted)

 

  • Catalog No:
  • ECHLCUBE6001
  • ECHLCUBE6014
  • ECHLCUBE6013
  • ECHLCUBE6011
  • 630-3325
  • ECHLPHAS0001
  • ECHLCUBE6007
  • ECHLCUBE6005
  • ECHLCUBE6004
  • ECHLCUBE6006
  • ECHLCUBE6003
  • ECHLCUBE6002
  • ECHLCUBE6010
  • ECHLCUBE6009
  • ECHLCUBE6008
  • Observation method:
  • Brightfield, phase contrast and fluorescence
  • Camera:
  • 12 MP colour (Brightfield), 5 MP monochrome camera (fluorescence)
  • Sensor:
  • CMOS
  • Display:
  • 12,9" iPad® Retina® display, 2732×2048 (264 pixels/inch)
  • Output:
  • USB
  • Electrical:
  • 100 - 240 VA, (50/60 Hz)

 

Objectives (by Olympus)

 

Frequently Bought Together