Orders LinkContactUsLinkComponent
sample_to_sequence_nap | Avantor
Nucleic Acid Purification
Mag.jpg
omega BIO-TEK
Omega Bio-tek Inc. offers high quality, convenient and cost-effective products for nucleic acid purification.
Learn more
Additional Resources
cfDNA kits
SeraXtractaDNAKit2.jpg
cytiva
Sera-Xtracta DNA kits based on magnetic beads: Sera-Xtracta HMW DNA Kit for high yield extraction of high-molecular-weight genomic DNA from blood, buffy coat, saliva, cultured mammalian cells and solid tissue. Sera-Xtracta Cell-Free DNA Kit for efficient extraction and purification of cfDNA from plasma.
cfDNA kit
Additional Resources
HMW genomic DNA kit
peqGOLD-.jpg
peqGOLD Nucleic Acid Purification kits. High quality silica based spin column kits for a wide range of sample types. Offering High yield, high purity and value for money
Plasmid Purification
gDNA purification
Additional Resources
RNA Purification