Order Entry
United States
Orders LinkContactUsLinkComponent
2,2'-Dithiodipyridine ≥99%

2,2'-Dithiodipyridine ≥99%

Plastic bottle, 1 kg
Catalog # CH00124-1KG
New Product
Supplier #:  00124-1KG
2,2'-Dithiodipyridine ≥99%
Catalog # CH00124-1KG
New Product
CAS Number:  2127-03-9
2,2'-Dithiodipyridine ≥99%
2,2'-Dithiodipyridine ≥99%

Plastic bottle, 1 kg

CAS Number:  2127-03-9
Price
$990.78
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

2,2'-Dithiodipyridine is a versatile compound widely recognized for its unique disulfide bond, which imparts significant stability and reactivity. This chemical is particularly valuable in the synthesis of various organic compounds, serving as a key intermediate in the production of pharmaceuticals, agrochemicals, and specialty chemicals. Its ability to form stable complexes with metal ions makes it an essential reagent in coordination chemistry, enhancing catalytic processes and facilitating the development of novel materials. Researchers often utilize 2,2'-Dithiodipyridine in the creation of functionalized polymers and as a protective agent in biochemical applications, showcasing its broad utility across multiple fields.

In addition to its synthetic applications, 2,2'-Dithiodipyridine is recognized for its role in the development of sensors and detection systems due to its electrochemical properties. Its unique structure allows for selective interactions with various analytes, making it an ideal candidate for innovative sensor technologies. With its combination of stability, reactivity, and versatility, 2,2'-Dithiodipyridine stands out as a crucial compound for researchers and industry professionals seeking to advance their projects in organic synthesis and material science.

Store at 0 to 8 °C.

Synonyms:  1,2-Di(pyridin-2-yl)disulphane, 1,2-Di(pyridin-2-yl)disulfane, 2,2'-Dithiodipyridine
  • CAS Number:2127-03-9
  • Formula:C₁₀H₈N₂S₂
  • MDL Number:MFCD00006287
  • Molecular Weight:220.23 g/mol
  • Storage Temperature:Refrigerator
  • UN:3335
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.