Order Entry
United States
Orders LinkContactUsLinkComponent
Bioran CPG 1000 A

Bioran CPG 1000 A

5 ml
Catalog # CH06992-5ML
New Product
Supplier #:  06992-5ML
Bioran CPG 1000 A
Catalog # CH06992-5ML
New Product
CAS Number:  
Bioran CPG 1000 A

5 ml

CAS Number:  
Price
$23.08
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Bioran CPG 1000 A is a biostimulant that enhances plant growth and resilience, optimizing crop yields while promoting soil health. Ideal for agriculture and research, it supports sustainable practices and innovative studies in plant physiology.

  • Store at room temperature

Specifications

Specifications

  • Size:
    5 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.