Order Entry
United States
Orders LinkContactUsLinkComponent
3-Methoxybenzhydrazide ≥99%

3-Methoxybenzhydrazide ≥99%

Plastic bottle, 250 g
Catalog # CH27124-250G
New Product
Supplier #:  27124-250G
3-Methoxybenzhydrazide ≥99%
Catalog # CH27124-250G
New Product
CAS Number:  5785-06-8
3-Methoxybenzhydrazide ≥99%
3-Methoxybenzhydrazide ≥99%

Plastic bottle, 250 g

CAS Number:  5785-06-8
Price
$383.76
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

3-Methoxybenzhydrazide is a versatile compound used in organic synthesis and pharmaceuticals. Its unique structure enables applications in agrochemicals, pharmaceuticals, and dye chemistry, making it a valuable reagent for researchers and industry professionals. Explore its potential in developing hydrazone derivatives and metal complexes.

Caution: Store at 0 to 8 °C.

Synonyms:  3-Methoxybenzohydrazide, 3-Methoxybenzene-1-carbohydrazide, m-Anisic hydrazide, 3-Anisohydrazide, 3-Methoxybenzhydrazide, 3-Methoxybenzoic acid hydrazide, 3-Methoxybenzoic hydrazide
  • CAS Number:5785-06-8
  • Formula:C₈H₁₀N₂O₂
  • MDL Number:MFCD00007601
  • Molecular Weight:166.18 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Purity:
    ≥99% (GC)
  • Apperance:
    Pale brown powder

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.