ANTIBODY ANTI-HUMAN IGG1+IGG2+ 100UL
Catalog # 78009-946
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-HUMAN IGG1+IGG2+ 100UL
Catalog # 78009-946
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322213-100
New Product
Frequently Bought Together
PI MARKER 5.85 200 UL
Supplier: 0000014126
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
PI MARKER 9.50 200UL
Supplier: 0000014126
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Potassium iodide
Supplier: G-Biosciences
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
Supplier: Revvity Health Sciences
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: 0000014126
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
72262 ALL TRANS RETINOIC ACID 50MG
Supplier: STEMCELL Technologies
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen