ASSAY KIT MDH (NAD+/NADH) 96TST
78009-890
New Product
Frequently Bought Together
Bovine Plasma with Citrate, Equitech- Bio, Inc.
: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
: Equitech-Bio
HEPES Buffer, 0.2M Buffer soln., pH 7.6, Low Endotoxin, Thermo Scientific
: Thermo Scientific Chemicals
ELISA KIT ALDOSTERONE CHEMILUMINESCENTS
: 0000018898
Rat Plasma, Equitech- Bio
: Equitech-Bio
DEFIBRINATED BLOOD SHEEP 100ML
: LAMPIRE BIOLOGICAL LAB MS
ELISA KIT EPIANDROSTERONES
: 0000018898
ELISA KIT CORTISOLS
: ARBOR ASSAYS MS
RIPA Lysis Buffer, Rockland Immunochemicals
: Rockland Immunochemical
Human IL-10 ELISA Kit, Rockland Immunochemicals
: Rockland Immunochemical
1M SOD ACETATE PH 1L STERILE
: Teknova
Fixation, (buffer), Enzo Life Sciences
: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
: Innovative Research Inc
Fixation/Permeabilization, (buffer), Enzo Life Sciences
: Enzo Life Sciences
Sodium Dodecyl Sulfate SDS, 20%
: Rockland Immunochemical
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
: Genetex
Goat Plasma, Equitech- Bio, Inc.
: Equitech-Bio
BUFFER JAW SSC 2X 1L PACKS
: G-Biosciences