ELISA KIT MOUSE BMP1/PCP A2375 48TEST
Catalog # 78008-546
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE BMP1/PCP A2375 48TEST
Catalog # 78008-546
Supplier: ANTIBODIES.COM LLC
Supplier Number: A2375-48
New Product
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
PI MARKER 9.50 200UL
Supplier: 0000014126
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
PI MARKER ICE280/ICE3 4.65 200UL
Supplier: 0000014126
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Radiance Q Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
DNA WASH BUFFER 6 ML
Supplier: Zymo Research