ELISA KIT HUMAN ABP1 A2202 48TEST
Catalog # 78008-242
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT HUMAN ABP1 A2202 48TEST
Catalog # 78008-242
Supplier: ANTIBODIES.COM LLC
Supplier Number: A2202-48
New Product
Frequently Bought Together
VWR® Stripping Buffers
Supplier: VWR
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Radiance Q Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
BUFFER LDS SAMPLE LOADING (4X) 10ML
Supplier: G-Biosciences