ASSAY KIT TANNIN A319743 96TEST
Catalog # 78000-299
Supplier: ANTIBODIES.COM LLC
New Product
ASSAY KIT TANNIN A319743 96TEST
Catalog # 78000-299
Supplier: ANTIBODIES.COM LLC
Supplier Number: A319743-96
New Product
Frequently Bought Together
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
10X Tris-Glycine
Supplier: Rockland Immunochemical
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
ELISA ARG8 VASOPRESSIN CHEMILUMINESCENTS
Supplier: 0000018898
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
g-DNA Wash Buffer, Zymo Research
Supplier: Zymo Research
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences