ASSAY PEPC ACTIVITY A319716 96TEST
Catalog # 78000-250
Supplier: ANTIBODIES.COM LLC
New Product
ASSAY PEPC ACTIVITY A319716 96TEST
Catalog # 78000-250
Supplier: ANTIBODIES.COM LLC
Supplier Number: A319716-96
New Product
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Western Blot Boxes
Supplier: Alkali Scientific
MARKER MAURICE CIEF PI-8.40 LYOPH 210UL
Supplier: 0000014126
PI MARKER 9.50 200UL
Supplier: 0000014126
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: 0000014126
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio