ASSAY HYDROGEN PEROXIDE A319692-96TEST
Catalog # 78000-217
Supplier: ANTIBODIES.COM LLC
New Product
ASSAY HYDROGEN PEROXIDE A319692-96TEST
Catalog # 78000-217
Supplier: ANTIBODIES.COM LLC
Supplier Number: A319692-96
New Product
Frequently Bought Together
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
MARKER 9.99 MAURICE CIEF PI 210 UL
Supplier: 0000014126
PI MARKER MAURICE CIEF 4.05 210UL
Supplier: 0000014126
PI MARKER 9.50 200UL
Supplier: 0000014126
Chemi Blot Blocking Buffer, Azure
Supplier: Azure Biosystems
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Buffer P3 (Yellow), Zymo Research
Supplier: Zymo Research
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Western Blot Boxes
Supplier: Alkali Scientific