pH Standard Buffers, Highly Acidic, Inorganic Ventures
Supplier: Inorganic Ventures
Supplier #: PH-3-1L
pH Standard Buffers, Highly Acidic, Inorganic Ventures
Catalog # 77999-395
Supplier: Inorganic Ventures
New Product
CAS Number:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
- Certified reference material set in a water matrix for stability.
- These standards are Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines.
- One year Expiration Date.
Documents
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



