EPA Method 200.8 Internal Standards
500 ml, 20 µg/ml each of Bismuth, Scandium, Yttrium, Indium, and Terbium, 5% HNO₃, LDPE bottle
Catalog # 77996-879
New Product
Supplier: Inorganic Ventures
Supplier #: 2008ISS-500ML
EPA Method 200.8 Internal Standards
Catalog # 77996-879
Supplier: Inorganic Ventures
New Product
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Certifications: ISO 17025/17034.
Specifications
Specifications
- Matrix:5% HNO₃
- Pack type:LDPE bottle
- Size:500 ml
- Elements and concentration:20 µg/ml each of Bismuth, Scandium, Yttrium, Indium, and Terbium
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



