ASSAY KIT CHITOSAN 96 WELL
Catalog # 104091-034
Supplier: Cell Biolabs
Frequently Bought Together
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
100 1042 CHIR99021
Supplier: STEMCELL Technologies
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
BUFFER STABILIZR 7.5% DTPA-PURI BSA 50ML
Supplier: Revvity Health Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
100 0544 ERASTIN
Supplier: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Human Recombinant Shh (C24II)
Supplier: STEMCELL Technologies
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
100 0249 FORSKOLIN 25MG
Supplier: STEMCELL Technologies