ELISA KIT S-ADENOSYLMETHIONINE SAM
Catalog # 104090-968
Supplier: Cell Biolabs
ELISA KIT S-ADENOSYLMETHIONINE SAM
Catalog # 104090-968
Supplier: Cell Biolabs
Supplier Number: MET-5152
Frequently Bought Together
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
100 1042 CHIR99021
Supplier: STEMCELL Technologies
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Human Recombinant Shh (C24II)
Supplier: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
AGONIST RESINIFERATOXIN TRPV1 100UG
Supplier: FOCUS BIOMOLECULES MS
100 0249 FORSKOLIN 25MG
Supplier: STEMCELL Technologies
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
72262 ALL TRANS RETINOIC ACID 50MG
Supplier: STEMCELL Technologies
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
Supplier: Invitrogen
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences