MOUSE IGEA MATCHED PAIR A270517-576 TEST
Catalog # 77998-175
Supplier: ANTIBODIES.COM LLC
New Product
MOUSE IGEA MATCHED PAIR A270517-576 TEST
Catalog # 77998-175
Supplier: ANTIBODIES.COM LLC
Supplier Number: A270517-696
New Product
Frequently Bought Together
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
74042 PHORBOL12 MYRISTATE 13 ACETATE 1MG
Supplier: STEMCELL Technologies
100 1042 CHIR99021
Supplier: STEMCELL Technologies
72192 PROSTAGLANDIN E2 1MG
Supplier: STEMCELL Technologies
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Human Recombinant Shh (C24II)
Supplier: STEMCELL Technologies
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
Supplier: Invitrogen
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS