ANTIBODY ANTI-IL-8 A269878 250UG
77997-599
New Product
Frequently Bought Together
Nano-Glo® Luciferase Assay System, Promega, Promega
: Promega Corporation
Bovine Plasma with Citrate, Equitech- Bio, Inc.
: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
: Equitech-Bio
Fixation, (buffer), Enzo Life Sciences
: Enzo Life Sciences
72192 PROSTAGLANDIN E2 1MG
: STEMCELL Technologies
KIT PCR ASSAY ELISA HU PAPPA -20C 2PLATE
: RAYBIOTECH LIFE INC MS
DEFIBRINATED BLOOD SHEEP 100ML
: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
: MP Biomedicals
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
: Invitrogen
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
: Genetex
AGONIST RESINIFERATOXIN TRPV1 100UG
: FOCUS BIOMOLECULES MS
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
: Invitrogen
100 0544 ERASTIN
: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
: Meso Scale Diagnostics
BUFFER JAW SSC 2X 1L PACKS
: G-Biosciences
BLOCKER AQB013 AQUAPORIN 5MG
: FOCUS BIOMOLECULES MS
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
: Revvity Health Sciences
Fixation/Permeabilization, (buffer), Enzo Life Sciences
: Enzo Life Sciences
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
: Invitrogen