MONKEY IL2 MATCHED PAIR A270512-576 TEST
Catalog # 77998-167
Supplier: ANTIBODIES.COM LLC
New Product
MONKEY IL2 MATCHED PAIR A270512-576 TEST
Catalog # 77998-167
Supplier: ANTIBODIES.COM LLC
Supplier Number: A270512-696
New Product
Frequently Bought Together
73884 DIBUTYRYL CAMP SODIUM SALT 100MG
Supplier: STEMCELL Technologies
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
72192 PROSTAGLANDIN E2 1MG
Supplier: STEMCELL Technologies
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
BLOCKER AQB013 AQUAPORIN 5MG
Supplier: FOCUS BIOMOLECULES MS
100 0251 ELTROMBOPAG 5MG
Supplier: STEMCELL Technologies
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
74042 PHORBOL12 MYRISTATE 13 ACETATE 1MG
Supplier: STEMCELL Technologies
100 1042 CHIR99021
Supplier: STEMCELL Technologies
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
SOLUTION ATTUNE WASH 250ML
Supplier: Invitrogen
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
Supplier: Invitrogen
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
BUFFER STABILIZR 7.5% DTPA-PURI BSA 50ML
Supplier: Revvity Health Sciences