Eisco Latex Dropper Bulbs
Catalog # 470344-007
Supplier: Wards
New Product
Specifications
- Ø:0.5"
- Capacity:10 ml
- Length:1.8"
Specifications
About this item
Bulbs made from high-quality latex fit pipettes and medicine droppers measuring 6 to 8 mm in diameter.
- Pack of 72 dropper bulbs made of high quality latex rubber
- Excellent for science classrooms and professional laboratories
Latex dropper bulbs fit glass pipettes.
Frequently Bought Together