ANTIGEN ANTHRAX PROACTIVE F/B ANTHRACIS
Catalog # MSPP-171E
Supplier: List Biological Laboratories, Inc.
ANTIGEN ANTHRAX PROACTIVE F/B ANTHRACIS
Catalog # MSPP-171E
Supplier: List Biological Laboratories, Inc.
Supplier Number: 171E
Frequently Bought Together
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
VWR® Acrylamide and Bis-Acrylamide
Supplier: VWR
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Glycine
Supplier: G-Biosciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
Upright Stainless Steel Freezer Rack for 2" Boxes, 4×5 Configuration
Supplier: Alkali Scientific
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
DNA WASH BUFFER 6 ML
Supplier: Zymo Research
Human/Mouse Recombinant Activin A
Supplier: STEMCELL Technologies
Yeast Mating Factor alpha (from Yeast)
Supplier: Genscript
Western Blot Boxes
Supplier: Alkali Scientific
Blue Prestained Protein Standard, Broad Range (11 to 250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Buffer P3 (Yellow), Zymo Research
Supplier: Zymo Research
DNA BINDING BUFFER 100 ML
Supplier: Zymo Research
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Chemi Blot Blocking Buffer, Azure
Supplier: Azure Biosystems
Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)