Order Entry
United States
Orders LinkContactUsLinkComponent
FUJIFILM Wako Zeranol Standard

FUJIFILM Wako Zeranol Standard

Zeranol standard for HPLC, 200 mg, ≥99.0% (HPLC), ≥98.0% (qNMR), Standard, Solid (powder/crystal), no matrix additives, Glass container, 1, N/A, Loss on drying ≤0.2%, Melting point, specific rotation, assay, solubility, HPLC assay ≥99.0%, qNMR assay ≥98.0%
Catalog # 77974-793
New Product
Supplier #:  262-01461
FUJIFILM Wako Zeranol Standard
Catalog # 77974-793
New Product
CAS Number:  
FUJIFILM Wako Zeranol Standard

Zeranol standard for HPLC, 200 mg, ≥99.0% (HPLC), ≥98.0% (qNMR), Standard, Solid (powder/crystal), no matrix additives, Glass container, 1, N/A, Loss on drying ≤0.2%, Melting point, specific rotation, assay, solubility, HPLC assay ≥99.0%, qNMR assay ≥98.0%

CAS Number:  
Price
$139.29
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Reagent for pesticide residue analysis.

  • High purity (HPLC ≥99.0%)
  • Reliable reference for Zeranol analysis
  • Suitable for regulatory and research applications
  • Consistent quality and validated specifications

Zeranol is a nonsteroidal estrogenic compound derived from zearalenone and widely studied for its occurrence in food and feed. FUJIFILM Wako’s Zeranol Standard is manufactured to meet the strict demands of HPLC analysis, ensuring consistent and accurate results in detection and quantification workflows. The material is supplied in a light-protected glass vial and should be stored refrigerated to maintain stability.

Zeranol ≥99.0% (HPLC), ≥98.0% (qNMR).

Specifications

Specifications

  • Concentration:
    ≥99.0% (HPLC), ≥98.0% (qNMR)
  • Matrix:
    Solid (powder/crystal), no matrix additives
  • Pack type:
    Glass container
  • Description:
    Zeranol standard for HPLC
  • Size:
    200 mg
  • No. of analytes:
    1
  • Shelf life:
    N/A
  • Accuracy:
    HPLC assay ≥99.0%, qNMR assay ≥98.0%
  • Certification:
    SDS and specification document available
  • Tolerance:
    Loss on drying ≤0.2%
  • Type:
    Standard
  • Parameters:
    Melting point, specific rotation, assay, solubility
  • Cat. No.:
    77974-793
  • Formula:
    C₁₈H₂₆O₅
  • Instrument:
    HPLC
  • Physical Form:
    White to nearly white crystals or powder
  • Method:
    HPLC, qNMR
  • Shipping Conditions:
    RT
  • Formula Weight:
    322.39
  • CAS No.:
    26538-44-3
  • Storage Temperature:
    Refrigerated (storage at 2...10 °C)

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.