Solution Bottles, Sterile
Solution bottle, sterile, 500 ml, Polystyrene, Clear
Catalog # 77973-186
New Product
Supplier: CELLTREAT Scientific Products
Supplier #: 229784
Jump to:
Product Details & Documents
Solution bottles are key for storage, preparation, and dispensing of chemical solutions and reagents.
- Compatible with bottle top filters or for use as solution storage containers
Solution bottle neck size is 45 mm
Curved bottle design for easy handling
Specifications
- Capacity:500 ml
- Color:Clear
- Description:Solution bottle, sterile
- Material:Polystyrene



